Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ABCA1 protein, human recombinant | >90.0% | 10UG
SDP

Catalog No. 5000175807
Encompass_Preferred

Recombinant human ABCA1 protein produced in HEK293 cells with N-StrepII, C-FLAG, and N-8xHis affinity tags. Designed for research applications such as studies of lipid transport, cholesterol efflux, and HDL biogenesis. Characterized by reducing SDS-PAGE and supplied in microgram-scale quantities.

  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Multiple affinity tags to facilitate purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 258.4 kDa.
  • Supplied in small microgram quantities suitable for analytical and functional assays.

Catalog No. 50-001-75807 Supplier Medchemexpress LLC Supplier No. HYP70135710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human ABCA1 protein produced in HEK293 cells with N-StrepII, C-FLAG, and N-8xHis affinity tags. Designed for research applications such as studies of lipid transport, cholesterol efflux, and HDL biogenesis. Characterized by reducing SDS-PAGE and supplied in microgram-scale quantities.

  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Multiple affinity tags to facilitate purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 258.4 kDa.
  • Supplied in small microgram quantities suitable for analytical and functional assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.