Learn More
Aviva Systems Biology Corp ABCA12 Antibody - middle region (ARP43683_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43683_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCA12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ABCA12. Alternate names: DKFZp434G232, FLJ41584, ICR2B, LI2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58605