Learn More
Aviva Systems Biology Corp ABCA5 Antibody - C-terminal region (ARP43688_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43688_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABCA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ABCA5. Alternate names: ABC13, DKFZp451F117, DKFZp779N2435, EST90625, FLJ16381ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58607