Learn More
Aviva Systems Biology Corp Abca7 Antibody - middle region (ARP43690_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43690_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Abca7. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: ABCX, Abc51ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-285-092