Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Alexa Fluor™ 700 Conjugation Kit (Fast) - Lightning-Link™, 3X100UG

Catalog No. NC2151223
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC2151223 3 x 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NC2151223 Supplier Abcam Supplier No. AB2698243X100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, with 4 citations.

Specifications

Product Type Secondary Antibody
Shipping Condition Blue Ice
Conjugate Alexa Fluor 700
Quantity 3 x 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.