Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam all-cis-7,10,13,16,19-Docosapentaenoic acid, Long chain polyunsaturated fatty acid, 25 mg

Catalog No. 502658084
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-8084 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-8084 Supplier Abcam Supplier No. AB14389425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody.

  • Storage Buffer: Constituents: 99% Ethanol, 1% all-cis-7,10,13,16,19-Docosapentaenoic ac
  • Form: Liquid

Specifications

Quantity 25 mg
Formulation Constituents: 99% Ethanol, 1% all-cis-7,10,13,16,19-Docosapentaenoic ac
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.