Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Anti-Human IgG4 pFc' [HP6023] (FITC), 250 ug

Catalog No. 502648667
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-264-8667 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-264-8667 Supplier Abcam Supplier No. AB99815250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal antibody, suitable for Dot, ELISA, WB, ICC/IF, with 2 citations.

  • Host Species: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG3
  • Applications: Dot, ELISA, WB, ICC/IF
  • Species Reactivity: Human
  • Storage Buffer: pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification Protein A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications Dot Blot, ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone HP6023
Conjugate Unconjugated
Formulation pH: 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS
Gene IGHG4
Gene Symbols IGHG4
Host Species Mouse
Purification Method Affinity purification Protein A
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Human.
Target Species Human
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG3
Host abrv Ms
Shipping Condition Blue Ice
Reactivity abrv Hu
Target Molecule IGHG4, Human
Validated Application abrv Dot, ELISA, WB, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.