Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Anti-mouse IgG for IP (Biotin), 50 ug

Catalog No. NC3135928
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC3135928 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC3135928 Supplier Abcam Supplier No. AB13136750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal secondary antibody, suitable for WB, with 2 citations.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: WB
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.09% Sodium azide, Constituents: 0.88% Sodium chloride, 0.16% Sodium phosphate
  • Form: Liquid

Specifications

Applications Western Blot
Classification Monoclonal
Clone eB144
Conjugate Biotin
Formulation Preservative: 0.09% Sodium azide, Constituents: 0.88% Sodium chloride, 0.16% Sodium phosphate
Gene Symbols Ighg1
Host Species Rat
Quantity 50 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.