Learn More
Abcam Anti-mouse IgG for IP (HRP), 100 uL
Description
- Host Species: Rat
- Clonality: Monoclonal
- Isotype: IgG
- Applications: WB
- Species Reactivity: Mouse
- Storage Buffer: Constituents: 50% Glycerol (glycerin, glycerine), 0.88% Sodium chloride, 0.16% Sodium phosphate, 0.1% BSA
- Form: Liquid
- Purification: Affinity purification Protein G
Specifications
Specifications
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | eB144 |
| Conjugate | HRP |
| Formulation | Constituents: 50% Glycerol (glycerin, glycerine), 0.88% Sodium chloride, 0.16% Sodium phosphate, 0.1% BSA |
| Gene Symbols | Ighg1 |
| Host Species | Rat |
| Purification Method | Affinity purification Protein G |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.