Learn More
Abcam Anti-STING antibody [EPR25090-107], 100 uL
Description
- Host Species: Rabbit
- Clonality: Monoclonal
- Isotype: IgG
- Applications: Flow Cyt (Intra), IP, WB, IHC-P, IHC-Fr, ICC/IF
- Species Reactivity: Rat and Mouse
- Storage Buffer: pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
- Form: Liquid
- Purification: Affinity purification Protein A
Specifications
Specifications
| Applications | Flow Cytometry (Intracellular Staining), Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | EPR25090-107 |
| Conjugate | Unconjugated |
| Formulation | pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA |
| Gene Symbols | Sting1 |
| Host Species | Rabbit |
| Purification Method | Affinity purification Protein A |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.