Learn More
Abcam Arachidonic acid, omega-6 Polyunsaturated fatty acid, 250 mg
Description
- Storage Buffer: Constituents: 75% Ethanol, 25% Arachidonic acid
- Form: Liquid
Specifications
Specifications
| Quantity | 250 μg |
| Formulation | Constituents: 75% Ethanol, 25% Arachidonic acid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.