Learn More
Abcam Autophagy Marker (APG5L/ATG5, ATG16L1, ATG4B, ATG9A, Beclin 1, LC3B) Antibody Sampler Panel, 1 Kit
Description
- Applications: WB
- Species Reactivity: Mouse, Rat and Human
Specifications
Specifications
| Applications | Western Blot |
| Conjugate | Unconjugated |
| Gene Symbols | ATG5 |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Test Specificity | This product reacts with Mouse, Rat and Human. |
| Target Species | Mouse, Rat, Human |
| Product Type | Secondary Antibody |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.