Learn More
Abcam Biotinylated Goat Anti-Rabbit IgG (H+L) (Ready to use), 125 mL
Description
- Host Species: Goat
- Clonality: Polyclonal
- Applications: IHC-P
- Storage Buffer: pH: 7.2 - 7.6, Preservative: 0.0985% Sodium azide, Constituents: PBS
- Form: Liquid
Specifications
Specifications
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | pH: 7.2 - 7.6, Preservative: 0.0985% Sodium azide, Constituents: PBS |
| Host Species | Goat |
| Quantity | 125 mL |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Packaging | Vial |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.