Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Caspase 3/7 Staining Kit (Far Red), 50 Tests

Catalog No. NC2403529
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC2403529 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC2403529 Supplier Abcam Supplier No. AB27078550TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody, suitable for Flow Cyt, Fluorescence Microscopy, Enzyme activity assay, with 2 citations.

  • Applications: Flow Cyt, Fluorescence Microscopy, Enzyme activity assay

Specifications

Product Type Secondary Antibody
Shipping Condition Blue Ice
Conjugate Unconjugated
Quantity 50 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.