Learn More
Abcam Chicken Anti-Mouse IgG H&L (TRITC), 1 mg
Description
- Host Species: Chicken
- Clonality: Polyclonal
- Applications: Flow Cyt, IHC-Fr, IHC-P, IM, ICC/IF, ELISA
- Species Reactivity: Mouse
- Storage Buffer: Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
- Form: Liquid
- Purification: Affinity purification Immunogen
Specifications
Specifications
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | TRITC |
| Formulation | Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution |
| Gene Symbols | Ighg1 |
| Host Species | Chicken |
| Purification Method | Affinity purification Immunogen |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.