Learn More
Abcam Chicken Anti-Rabbit IgG H&L (HRP), 1 mg
Description
- Host Species: Chicken
- Clonality: Polyclonal
- Isotype: IgG
- Applications: Dot, IHC-Fr, IHC-P, IM, ICC, ELISA, WB
- Species Reactivity: Rabbit
- Storage Buffer: Preservative: 0.01% Gentamicin sulphate, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
- Form: Liquid
- Purification: Affinity purification
Specifications
Specifications
| Applications | Dot Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry, ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | Preservative: 0.01% Gentamicin sulphate, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution |
| Host Species | Chicken |
| Purification Method | Affinity purification |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.