Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam cis-6-Hexadecenoic acid, Monounsaturated fatty acid, 25 mg

Catalog No. 502658083
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-8083 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-8083 Supplier Abcam Supplier No. AB14391125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibody.

  • Storage Buffer: Constituents: 100% (Z)-hexadec-6-enoic acid
  • Form: Liquid

Specifications

Quantity 25 mg
Formulation Constituents: 100% (Z)-hexadec-6-enoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.