Learn More
Abcam Cytochrome c Apoptosis ICC Antibody Kit, 200 ug
Description
- Applications: ICC/IF
- Species Reactivity: Mouse, Rat and Human
Specifications
Specifications
| Applications | Immunocytochemistry, Immunofluorescence |
| Conjugate | Unconjugated |
| Gene Symbols | CYCS |
| Quantity | 200 μg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Test Specificity | This product reacts with Mouse, Rat and Human. |
| Target Species | Mouse, Rat, Human |
| Product Type | Secondary Antibody |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.