Learn More
Abcam Donkey Anti-Chicken IgY H&L (FITC), 500 ug
Description
- Host Species: Donkey
- Clonality: Polyclonal
- Isotype: IgG
- Applications: IHC-Fr
- Species Reactivity: Chicken
- Storage Buffer: pH: 7.2, Preservative: 0.01% Thimerosal (merthiolate), Constituents: PBS, 50% Glycerol (glycerin, glycerine)
- Form: Liquid
- Purification: Affinity purification Protein L
Specifications
Specifications
| Applications | Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | pH: 7.2, Preservative: 0.01% Thimerosal (merthiolate), Constituents: PBS, 50% Glycerol (glycerin, glycerine) |
| Host Species | Donkey |
| Purification Method | Affinity purification Protein L |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.