Learn More
Abcam Donkey Anti-Goat IgG H&L (Alexa Fluor™ 594) preadsorbed, 500 ug
Description
- Host Species: Donkey
- Clonality: Polyclonal
- Isotype: IgG
- Applications: ELISA, Flow Cyt, ICC/IF, IHC-Fr, IHC-P
- Species Reactivity: Goat
- Storage Buffer: PBS
- Form: Liquid
- Purification: Affinity purification Immunogen
Specifications
Specifications
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA |
| Host Species | Donkey |
| Purification Method | Affinity purification Immunogen |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.