Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Donkey Anti-Goat IgG H&L (DyLight™ 488) preadsorbed, 500 ug

Catalog No. 502648854
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-264-8854 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-264-8854 Supplier Abcam Supplier No. AB96935500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Donkey Polyclonal secondary antibody, suitable for ICC/IF, Flow Cyt, IHC-P, WB, with 9 citations.

  • Host Species: Donkey
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ICC/IF, Flow Cyt, IHC-P, WB
  • Species Reactivity: Goat
  • Storage Buffer: pH: 6.8 - 7.4, Preservative: 0.09% Sodium azide, Constituents: PBS, 0.2% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 6.8 - 7.4, Preservative: 0.09% Sodium azide, Constituents: PBS, 0.2% BSA
Host Species Donkey
Purification Method Affinity purification Immunogen
Quantity 500 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Goat.
Target Species Goat
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Reactivity abrv Gt
Validated Application abrv ICC/IF, Flow Cyt, IHC-P, WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.