Learn More
Abcam Donkey Anti-Goat IgG H&L (HRP polymer), prediluted, 15 mL
Description
- Host Species: Donkey
- Clonality: Polyclonal
- Isotype: IgG
- Applications: Ie, IHC-P
- Form: Liquid
Specifications
Specifications
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Donkey |
| Quantity | 15 mL |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Packaging | Vial |
| Target Species | Non-species specific |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.