Learn More
Abcam Donkey Anti-Mouse IgG H&L (10nm Gold), 1250 uL
Description
- Host Species: Donkey
- Clonality: Polyclonal
- Isotype: IgG
- Applications: EM
- Species Reactivity: Mouse
- Storage Buffer: Preservative: 0.097% Sodium azide, Constituents: PBS, 1% BSA
- Form: Liquid
- Purification: Affinity purification Immunogen
Specifications
Specifications
| Applications | Electron Microscopy |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Preservative: 0.097% Sodium azide, Constituents: PBS, 1% BSA |
| Gene Symbols | Ighg1 |
| Host Species | Donkey |
| Purification Method | Affinity purification Immunogen |
| Quantity | 1250 μL |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.