Learn More
Abcam Donkey Anti-Mouse IgG H&L (Biotin), 500 ug
Description
- Host Species: Donkey
- Clonality: Polyclonal
- Isotype: IgG
- Applications: ELISA, Flow Cyt, ICC/IF, IHC-Fr, IHC-P, IP, WB
- Species Reactivity: Mouse
- Storage Buffer: pH: 7.4, Preservative: 0.02% Sodium azide, Constituents: PBS, 30% Glycerol (glycerin, glycerine), 1% BSA
- Form: Liquid
- Purification: Affinity purification
Specifications
Specifications
| Applications | Western Blot, Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | pH: 7.4, Preservative: 0.02% Sodium azide, Constituents: PBS, 30% Glycerol (glycerin, glycerine), 1% BSA |
| Gene Symbols | Ighg1 |
| Host Species | Donkey |
| Purification Method | Affinity purification |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.