Learn More
Abcam Donkey Anti-Mouse IgG H&L (PE) preadsorbed, 250 ug
Description
- Host Species: Donkey
- Clonality: Polyclonal
- Isotype: IgG
- Applications: Dot, Flow Cyt, ICC/IF, IHC-Fr, IHC-P, IM
- Species Reactivity: Mouse
- Storage Buffer: BSA, Potassium phosphate, Sodium chloride
- Form: Liquid
- Purification: Affinity purification
Specifications
Specifications
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry, Dot Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution |
| Gene Symbols | Ighg1 |
| Host Species | Donkey |
| Purification Method | Affinity purification |
| Quantity | 250 μg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.