Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Donkey Anti-Rabbit IgG H&L (10nm Gold), 1250 uL

Catalog No. 502650817
Encompass_Preferred
Change view
Click to view available options
Quantity:
1250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-0817 1250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-0817 Supplier Abcam Supplier No. AB395971250UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Donkey Polyclonal secondary antibody, suitable for EM, with 4 citations.

  • Host Species: Donkey
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: EM
  • Species Reactivity: Rabbit
  • Storage Buffer: Preservative: 0.097% Sodium azide, Constituents: PBS, 1% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications Electron Microscopy
Classification Polyclonal
Conjugate Unconjugated
Formulation Preservative: 0.097% Sodium azide, Constituents: PBS, 1% BSA
Host Species Donkey
Purification Method Affinity purification Immunogen
Quantity 1250 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Rabbit.
Target Species Rabbit
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Reactivity abrv Rb
Validated Application abrv EM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.