Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Donkey Anti-Rat IgG H&L (HRP), 1 mg

Catalog No. 502620781
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-262-0781 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-262-0781 Supplier Abcam Supplier No. AB1021821MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Donkey Polyclonal secondary antibody, suitable for IHC-P, ICC, WB, ELISA, with 8 citations.

  • Host Species: Donkey
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: IHC-P, ICC, WB, ELISA
  • Species Reactivity: Rat
  • Storage Buffer: pH: 6.8 - 7.4, Preservative: 0.05% CMIT/MIT based preservative, Constituents: 0.2% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation pH: 6.8 - 7.4, Preservative: 0.05% CMIT/MIT based preservative, Constituents: 0.2% BSA
Host Species Donkey
Purification Method Affinity purification Immunogen
Quantity 1 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Rat.
Target Species Rat
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Reactivity abrv Rt
Validated Application abrv IHC-P, ICC, WB, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.