Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Donkey Anti-Sheep IgG H&L (Biotin), 1 mg

Catalog No. 502649973
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-264-9973 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-264-9973 Supplier Abcam Supplier No. AB68991MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Donkey Anti-Sheep IgG H&L (Biotin)

  • Host Species: Donkey
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: Dot, ELISA, ICC/IF, IHC-Fr, IHC-P, IM, WB
  • Species Reactivity: Sheep
  • Storage Buffer: BSA, Potassium phosphate, Sodium chloride
  • Form: Liquid
  • Purification: Affinity purification

Specifications

Applications Dot Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), ELISA, Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Host Species Donkey
Purification Method Affinity purification
Quantity 1 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Sheep.
Target Species Sheep
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Reactivity abrv Sh
Validated Application abrv Dot, IM, IHC-Fr, IHC-P, ELISA, WB, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.