Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Fraction-PREP Cell Fractionation Kit, 50 Tests

Catalog No. 502205946
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-5946 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-5946 Supplier Abcam Supplier No. AB28808550TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant protein, with 2 citations.

Specifications

Name Fraction-PREP Cell Fractionation Kit
Quantity 50 Tests
Storage Requirements Please refer to protocols
Regulatory Status RUO
Product Type Recombinant Protein
Conjugate Unconjugated
Content And Storage Please refer to protocols
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.