Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Goat Anti-Armenian hamster IgG H&L (Texas Red ™), 1 mg

Catalog No. 502659175
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-9175 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-9175 Supplier Abcam Supplier No. AB57431MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Anti-Armenian hamster IgG H&L (Texas Red)

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ELISA, Flow Cyt, ICC/IF, IM, WB
  • Species Reactivity: Armenian hamster
  • Storage Buffer: BSA, Potassium phosphate, Sodium chloride
  • Form: Liquid
  • Purification: Affinity purification

Specifications

Applications Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry, ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Host Species Goat
Purification Method Affinity purification
Quantity 1 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Armenian hamster.
Target Species Armenian Hamster
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Validated Application abrv WB, IM, ICC/IF, Flow Cyt, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.