Learn More
Abcam Goat Anti-Chicken IgY H&L (25nm Gold), 1750 uL
Description
- Host Species: Goat
- Clonality: Polyclonal
- Isotype: IgG
- Applications: EM
- Species Reactivity: Chicken
- Storage Buffer: Preservative: 0.097% Sodium azide, Constituents: PBS, 1% BSA
- Form: Liquid
- Purification: Affinity purification Immunogen
Specifications
Specifications
| Applications | Electron Microscopy |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Preservative: 0.097% Sodium azide, Constituents: PBS, 1% BSA |
| Host Species | Goat |
| Purification Method | Affinity purification Immunogen |
| Quantity | 1750 μL |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.