Learn More
Abcam Goat Anti-Chicken IgY H&L (Biotin), 1 mg
Description
- Host Species: Goat
- Clonality: Polyclonal
- Isotype: IgG
- Applications: Dot, ELISA, ICC/IF, IHC-Fr, IHC-P, IM, WB
- Species Reactivity: Chicken
- Storage Buffer: BSA, Sodium chloride, Sodium phosphate
- Form: Liquid
- Purification: Affinity purification
Specifications
Specifications
| Applications | Dot Blot, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution |
| Host Species | Goat |
| Purification Method | Affinity purification |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.