Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Goat Anti-Chicken IgY H&L (PE), 250 ug

Catalog No. 502674073
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-267-4073 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-267-4073 Supplier Abcam Supplier No. AB46852250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal secondary antibody, suitable for WB, Dot, ELISA, ICC/IF.

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: WB, Dot, ELISA, ICC/IF
  • Species Reactivity: Chicken
  • Storage Buffer: pH: 7.3, Preservative: 0.09% Sodium azide, Constituents: PBS, 16% Sucrose
  • Form: Liquid

Specifications

Applications Western Blot, Dot Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate PE
Formulation pH: 7.3, Preservative: 0.09% Sodium azide, Constituents: PBS, 16% Sucrose
Host Species Goat
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Chicken.
Target Species Chicken
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Reactivity abrv Ck
Validated Application abrv WB, Dot, ELISA, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.