Learn More
Abcam Goat Anti-Guinea pig IgG H&L (Cy3.5 ™) preadsorbed, 500 ug
Description
- Host Species: Goat
- Clonality: Polyclonal
- Applications: ELISA, Flow Cyt, ICC, ICC/IF, IHC-Fr, IHC-P
- Species Reactivity: Guinea pig
- Storage Buffer: BSA, Potassium phosphate, Sodium chloride
- Form: Liquid
- Purification: Affinity purification Immunogen
Specifications
Specifications
| Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | pH: 6.5, Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.87% Sodium chloride, 0.42% Tripotassium orthophosphate |
| Host Species | Goat |
| Purification Method | Affinity purification Immunogen |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.