Learn More
Abcam Goat Anti-Human IgG+IgM+IgA H&L (Biotin), 1 mg
Description
- Species Reactivity: Human
- Storage: Avoid freeze / thaw cycle
Specifications
Specifications
| Antigen | Q6GMX6 |
| Applications | Immunohistochemistry |
| Gene Alias | Immunoglobulin heavy constant gamma 1, Ig gamma-1 chain C region, Ig gamma-1 chain C region EU, Ig gamma-1 chain C region KOL, Ig gamma-1 chain C region NIE, IGHG1 |
| Gene Symbols | IGHG1 |
| Quantity | 1 mg |
| Gene ID (Entrez) | 3507 |
| Target Species | Human |
| Content And Storage | Avoid freeze/thaw cycle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.