Learn More
Abcam Goat Anti-Mouse IgG (40nm Gold, 10 OD), 1 mL
Description
- Host Species: Goat
- Clonality: Polyclonal
- Isotype: IgG
- Applications: LFA
- Species Reactivity: Mouse
- Storage Buffer: pH: 7, Constituents: Tris buffered saline, 0.1% Sorbitan monolaurate, ethoxylated
- Form: Liquid
Specifications
Specifications
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | pH: 7, Constituents: Tris buffered saline, 0.1% Sorbitan monolaurate, ethoxylated |
| Gene Symbols | Ighg1 |
| Host Species | Goat |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Packaging | Vial |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.