Learn More
Abcam Goat Anti-Mouse IgG H&L (Alkaline Phosphatase) preadsorbed, 500 ug
Description
- Host Species: Goat
- Clonality: Polyclonal
- Applications: Dot, ELISA, ICC/IF, IHC-Fr, IHC-P, IM, WB
- Species Reactivity: Mouse
- Storage Buffer: BSA, Glycerol, Sodium chloride, Tris HCl, Magnesium chloride, Zinc chloride
- Form: Liquid
- Purification: Affinity purification
Specifications
Specifications
| Applications | Dot Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), ELISA, Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | pH: 8, Preservative: 0.09% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 1% BSA, 0.88% Sodium chloride, 0.788% Tris HCl, 0.0095% Magnesium chloride, 0.0014% Zinc chloride |
| Gene Symbols | Ighg1 |
| Host Species | Goat |
| Purification Method | Affinity purification |
| Quantity | 500 μg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.