Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Goat Anti-Mouse IgG H&L (FITC) preadsorbed, 500 ug

Catalog No. 502611679
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-261-1679 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-261-1679 Supplier Abcam Supplier No. AB7064500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Anti-Mouse IgG H&L (FITC) preadsorbed

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: Dot, ELISA, Flow Cyt, ICC/IF, IHC-Fr, IHC-P, IM, WB
  • Species Reactivity: Mouse
  • Storage Buffer: BSA, Potassium phosphate, Sodium chloride
  • Form: Liquid
  • Purification: Affinity purification

Specifications

Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Flow Cytometry, ELISA, Western Blot, Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Gene Symbols Ighg1
Host Species Goat
Purification Method Affinity purification
Quantity 500 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv IHC-Fr, IHC-P, IM, ICC/IF, Flow Cyt, ELISA, WB, Dot
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.