Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Goat Anti-Mouse IgG H&L (HRP polymer), 15 mL

Catalog No. 502588179
Encompass_Preferred
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-8179 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-8179 Supplier Abcam Supplier No. AB21487915ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal secondary antibody, suitable for Ie, IHC-P.

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: Ie, IHC-P
  • Form: Liquid

Specifications

Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Gene Symbols Ighg1
Host Species Goat
Quantity 15 mL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Target Species Non-species specific
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Validated Application abrv Ie, IHC-P
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.