Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Goat Anti-Mouse IgG1 H&L (PE), 100 ug

Catalog No. 502648650
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-264-8650 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-264-8650 Supplier Abcam Supplier No. AB99912100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal secondary antibody, suitable for Flow Cyt.

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: Flow Cyt
  • Species Reactivity: Mouse
  • Storage Buffer: pH: 7.4, Preservative: 0.05% Sodium azide, Constituents: 6.84% Sucrose, 1.64% Sodium phosphate, 0.87% Sodium chloride, 0.1% BSA
  • Form: Lyophilized
  • Purification: Affinity purification Protein G

Specifications

Applications Flow Cytometry
Classification Polyclonal
Conjugate PE
Formulation pH: 7.4, Preservative: 0.05% Sodium azide, Constituents: 6.84% Sucrose, 1.64% Sodium phosphate, 0.87% Sodium chloride, 0.1% BSA
Gene Symbols Ighg1
Host Species Goat
Purification Method Affinity purification Protein G
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Polyclonal Secondary Antibody
Form Lyophilized
Isotype IgG
Host abrv Gt
Shipping Condition Ambient
Reactivity abrv Ms
Validated Application abrv Flow Cyt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.