Learn More
Abcam Goat Anti-Mouse IgG2b H&L (PE), 100 ug
Description
- Host Species: Goat
- Clonality: Polyclonal
- Isotype: IgG2b
- Applications: Flow Cyt
- Species Reactivity: Mouse
- Storage Buffer: pH: 7.4, Preservative: 0.013% Sodium azide, Constituents: 1.71% Sucrose, 1.64% Sodium phosphate, 0.87% Sodium chloride, 0.1% BSA
- Form: Liquid
- Purification: Size-exclusion chromatography
Specifications
Specifications
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | PE |
| Formulation | pH: 7.4, Preservative: 0.013% Sodium azide, Constituents: 1.71% Sucrose, 1.64% Sodium phosphate, 0.87% Sodium chloride, 0.1% BSA |
| Host Species | Goat |
| Purification Method | Size-exclusion chromatography |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.