Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Goat Anti-Rabbit IgG H&L (HRP), 1 mg

Catalog No. NC1859429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC1859429 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC1859429 Supplier Abcam Supplier No. AB970511MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Anti-Rabbit IgG H&L (HRP)

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ELISA, ICC, IHC-P, WB
  • Species Reactivity: Rabbit
  • Storage Buffer: PBS, BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications Immunohistochemistry (Paraffin), Immunocytochemistry, ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation pH: 6.8 - 7.4, Constituents: PBS, 0.2% BSA, 0.05% CMIT/MIT based preservative
Host Species Goat
Purification Method Affinity purification Immunogen
Quantity 1 mg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Rabbit.
Target Species Rabbit
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Reactivity abrv Rb
Validated Application abrv IHC-P, ICC, ELISA, WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.