Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Goat Anti-Rat IgG Fc (DyLight™ 550) preadsorbed, 500 ug

Catalog No. 502708370
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-8370 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-8370 Supplier Abcam Supplier No. AB96972500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal secondary antibody, suitable for ICC/IF, Flow Cyt, IHC-P.

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ICC/IF, Flow Cyt, IHC-P
  • Species Reactivity: Rat
  • Storage Buffer: pH: 6.8 - 7.4, Preservative: 0.09% Sodium azide, Constituents: PBS, 0.2% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 6.8 - 7.4, Preservative: 0.09% Sodium azide, Constituents: PBS, 0.2% BSA
Host Species Goat
Purification Method Affinity purification Immunogen
Quantity 500 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Rat.
Target Species Rat
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Reactivity abrv Rt
Validated Application abrv ICC/IF, Flow Cyt, IHC-P
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.