Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Goat Anti-Syrian hamster IgG H&L (PE/Cy7™) preadsorbed, 125 ug

Catalog No. 502609274
Encompass_Preferred
Change view
Click to view available options
Quantity:
125 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-260-9274 125 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-260-9274 Supplier Abcam Supplier No. AB130807125UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Anti-Syrian hamster IgG H&L (PE/Cy7) preadsorbed

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: Dot, ELISA, Flow Cyt, ICC/IF, WB
  • Species Reactivity: Syrian hamster
  • Storage Buffer: pH: 7.1, Preservative: 0.1% Sodium azide, Constituents: Sucrose, PBS
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifications

Applications Western Blot, Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.1, Preservative: 0.1% Sodium azide, Constituents: Sucrose, PBS
Host Species Goat
Purification Method Affinity purification Immunogen
Quantity 125 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Syrian hamster.
Target Species Syrian Hamster
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Validated Application abrv WB, Dot, ELISA, Flow Cyt, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.