Learn More
Abcam Goat F(ab')2 Anti-Mouse IgG - (Fab)'2 (PE), pre-adsorbed, 500 uL
Description
- Host Species: Goat
- Clonality: Polyclonal
- Isotype: IgG
- Applications: Dot, Flow Cyt, ICC/IF, IM, WB
- Species Reactivity: Mouse
- Storage Buffer: BSA, Sodium chloride, Sodium phosphate
- Form: Liquid
- Purification: Affinity purification
Specifications
Specifications
| Applications | Western Blot, Dot Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution |
| Gene Symbols | Ighg1 |
| Host Species | Goat |
| Purification Method | Affinity purification |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.