Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Goat monoclonal [RMG05] Anti-Mouse IgG F(ab) (Biotin), 50 ug

Catalog No. 502639392
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-263-9392 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-263-9392 Supplier Abcam Supplier No. AB27218050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Monoclonal secondary antibody, suitable for ELISA.

  • Host Species: Goat
  • Clonality: Monoclonal
  • Isotype: IgG
  • Applications: ELISA
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.09% Sodium azide, Constituents: PBS, 50% Glycerol (glycerin, glycerine), 1% BSA
  • Form: Liquid
  • Purification: Affinity purification Protein G

Specifications

Applications ELISA
Classification Monoclonal
Clone RMG05
Conjugate Biotin
Formulation Preservative: 0.09% Sodium azide, Constituents: PBS, 50% Glycerol (glycerin, glycerine), 1% BSA
Gene Symbols Ighg1
Host Species Goat
Purification Method Affinity purification Protein G
Quantity 50 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Reactivity abrv Ms
Validated Application abrv ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.