Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Goat Whole Molecule Anti-Cow IgG H&L (HRP), 1 mL

Catalog No. 502649963
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-264-9963 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-264-9963 Supplier Abcam Supplier No. AB69341ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Whole Molecule Anti-Cow IgG H&L (HRP)

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: Dot, ELISA, ICC/IF, IHC-Fr, IHC-P, IM, WB
  • Species Reactivity: Bovine
  • Storage Buffer: Potassium phosphate, Sodium chloride, Polyethylene glycol
  • Form: Liquid

Specifications

Applications Dot Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate HRP
Formulation Constituents: 1.5% BSA, 1.46% Sodium chloride, 0.164% Sodium phosphate
Host Species Goat
Quantity 1 mL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Packaging Vial
Test Specificity This product reacts with Cow.
Target Species Bovine
Product Type Polyclonal Secondary Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Reactivity abrv Cow
Validated Application abrv Dot, ELISA, ICC/IF, IHC-Fr, IHC-P, IM, WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.