Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam HRP Anti-Rabbit IgG antibody, 500 uL

Catalog No. NC2184249
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC2184249 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC2184249 Supplier Abcam Supplier No. AB288151500UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal antibody, suitable for EIA, WB, IHC, with over 100 citations.

  • Host Species: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: EIA, WB, IHC
  • Species Reactivity: Rabbit
  • Storage Buffer: pH: 7.2, Preservative: 0.03% Proclin 300, Constituents: PBS, 30% Glycerol (glycerin, glycerine), 0.5% BSA, 0.15% EDTA
  • Form: Liquid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications EIA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation pH: 7.2, Preservative: 0.03% Proclin 300, Constituents: PBS, 30% Glycerol (glycerin, glycerine), 0.5% BSA, 0.15% EDTA
Host Species Goat
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Rabbit.
Target Species Rabbit
Product Type Polyclonal Antibody
Form Liquid
Isotype IgG
Host abrv Gt
Shipping Condition Blue Ice
Reactivity abrv Rb
Validated Application abrv EIA, WB, IHC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.