Learn More
Abcam ioSkeletal Myocytes - Human iPSC derived cells, 1 SMALL
Description
- Applications: HTS, RT-PCR, WB, ICC, FuncS
- Storage Buffer: Constituents: 10% Dimethylsulfoxide
- Form: Liquid
Specifications
Specifications
| Applications | Western Blot, Immunocytochemistry |
| Conjugate | Unconjugated |
| Formulation | Constituents: 10% Dimethylsulfoxide |
| Gene Symbols | ioSk line |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Packaging | Small Vial |
| Target Species | Non-species specific |
| Product Type | Secondary Antibody |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.