Learn More
Abcam Mesenchymal Stromal Cell Marker (CD44, CD45, CD90 CD29, CD105) Antibody Panel - Human, 1 UNIT
Specifications
Specifications
| Conjugate | Unconjugated |
| Quantity | 1 Ea. |
| Regulatory Status | RUO |
| Whole Molecule | Yes |
| Primary or Secondary | Secondary |
| Target Species | Non-species specific |
| Product Type | Secondary Antibody |
| Shipping Condition | Blue Ice |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.